Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502764971
Encompass_Preferred
The Grainger 16T318 is a 3/8 inch drive click torque wrench made by ACDelco. It has a torque range of 10 to 80 foot pounds and is accurate to within +/- 4% of the preset value. The wrench has a reversible ratchet head, measures 15 inches long, and weighs 2 pounds. It features a knurled grip handle, matte finish, and a scale showing the preset torque values. This click-type torque wrench emits an audible click when the preset torque is reached. It would be useful for precisely tightening bolts and nuts on automotive or industrial applications.
Catalog No. 50-276-4971 Supplier Grainger Supplier No. 16T318
The Grainger 16T318 is a 3/8 inch drive click torque wrench made by ACDelco. It has a torque range of 10 to 80 foot pounds and is accurate to within +/- 4% of the preset value. The wrench has a reversible ratchet head, measures 15 inches long, and weighs 2 pounds. It features a knurled grip handle, matte finish, and a scale showing the preset torque values. This click-type torque wrench emits an audible click when the preset torque is reached. It would be useful for precisely tightening bolts and nuts on automotive or industrial applications.