Learn More
Grainger Threading Tool 3/16 Carbide
Supplier:  Grainger 16T336
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/16 inch shank diameter and 60 degree threading point. It has an overall length of 4 inches and can cut unified coarse right-hand threads from 4-56 to 1/4-20 TPI. The tool features TiN coating for increased tool life and has an included angle of 60 degrees. It is made in the USA.
Catalog No. 50-276-4973