Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502764974
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 1/4 inch maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse right hand threads from 4-56 to 1/4-20. The overall tool length is 3 inches and it weighs 0.04 lbs. This single point threading tool is made in the USA out of C2 grade high speed steel for good wear resistance. It is designed for use on lathes, screw machines, and CNC lathes for both production threading and general purpose threading applications on materials like aluminum, brass, bronze, and plastics.
Catalog No. 50-276-4974 Supplier Grainger Supplier No. 16T331
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 1/4 inch maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse right hand threads from 4-56 to 1/4-20. The overall tool length is 3 inches and it weighs 0.04 lbs. This single point threading tool is made in the USA out of C2 grade high speed steel for good wear resistance. It is designed for use on lathes, screw machines, and CNC lathes for both production threading and general purpose threading applications on materials like aluminum, brass, bronze, and plastics.