Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765606
Encompass_Preferred
The Grainger Approved Threading Tool is a high-speed steel threading tool with a 60 degree threading point. It has a 1/2 inch shank diameter and is 6 inches overall length. The threading tool has an American National thread profile and can cut threads from 4 to 56 threads per inch. It is designed for use on lathes, screw machines, and automatic chucking machines for both right- and left-hand threading applications. The tool is made in the USA out of C2 grade high-speed steel for durability and has an unfinished shank.
Catalog No. 50-276-5606 Supplier Grainger Supplier No. 16T277
The Grainger Approved Threading Tool is a high-speed steel threading tool with a 60 degree threading point. It has a 1/2 inch shank diameter and is 6 inches overall length. The threading tool has an American National thread profile and can cut threads from 4 to 56 threads per inch. It is designed for use on lathes, screw machines, and automatic chucking machines for both right- and left-hand threading applications. The tool is made in the USA out of C2 grade high-speed steel for durability and has an unfinished shank.