Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765607
Encompass_Preferred
The Grainger 16T280 is a 16-ton air/hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/2 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for portability with two wheels and a handle. The bottle jack uses hydraulic oil and compressed air to lift loads up to 32,000 pounds with minimal effort. It meets ASME PALD standards and has a one year warranty. The 16T280 is recommended for use in auto repair shops, construction sites, and manufacturing facilities.
Catalog No. 50-276-5607 Supplier Grainger Supplier No. 16T280
The Grainger 16T280 is a 16-ton air/hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/2 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for portability with two wheels and a handle. The bottle jack uses hydraulic oil and compressed air to lift loads up to 32,000 pounds with minimal effort. It meets ASME PALD standards and has a one year warranty. The 16T280 is recommended for use in auto repair shops, construction sites, and manufacturing facilities.