Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765716
Encompass_Preferred
The Grainger 16T287 is a 16 ton capacity hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a 2-1/2 inch ram and a 15-1/2 inch handle for pumping. It is designed for use on hard level surfaces for lifting equipment, dies, molds and more. The jack's capacity is 32,000 pounds.
Catalog No. 50-276-5716 Supplier Grainger Supplier No. 16T287
The Grainger 16T287 is a 16 ton capacity hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a 2-1/2 inch ram and a 15-1/2 inch handle for pumping. It is designed for use on hard level surfaces for lifting equipment, dies, molds and more. The jack's capacity is 32,000 pounds.