Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765752
Encompass_Preferred
The Grainger 16T275 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-15 inches and a lowered height of 9.5 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two-piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and has an overload valve to prevent over-extending. This heavy duty jack is designed for shop use and farm equipment lifting. With proper care it will provide reliable service.
Catalog No. 50-276-5752 Supplier Grainger Supplier No. 16T275
The Grainger 16T275 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-15 inches and a lowered height of 9.5 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two-piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and has an overload valve to prevent over-extending. This heavy duty jack is designed for shop use and farm equipment lifting. With proper care it will provide reliable service.