Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765753
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can cut up to 8 threads per inch. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life and lubricity. The overall length is 3-1/2 inches. This threading tool is made in the USA by Micro 100 Tool Corp. It can machine a wide range of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The 3/16 inch shank allows use in a variety of tool holders.
Catalog No. 50-276-5753 Supplier Grainger Supplier No. 16T264
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can cut up to 8 threads per inch. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life and lubricity. The overall length is 3-1/2 inches. This threading tool is made in the USA by Micro 100 Tool Corp. It can machine a wide range of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The 3/16 inch shank allows use in a variety of tool holders.