Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765014
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified coarse right hand threads. The overall length is 3-1/2" and it has a TiN coating for increased tool life. This threading tool is good for production threading applications in common steels, cast iron, stainless steel, and non-ferrous materials. It provides accurate threading with optimized chip control.
Catalog No. 50-276-5014 Supplier Grainger Supplier No. 16T327
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified coarse right hand threads. The overall length is 3-1/2" and it has a TiN coating for increased tool life. This threading tool is good for production threading applications in common steels, cast iron, stainless steel, and non-ferrous materials. It provides accurate threading with optimized chip control.