Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502768359
Encompass_Preferred
The Grainger 16T153 is a single-phase, open drip-proof electric motor with a 1/3 horsepower rating. It has a 115/230 volt input and 1725 RPM speed. The motor is totally enclosed and fan cooled with a 48 frame size. It has a 1.15 service factor and is CSA certified. The 16T153 can be used for various light industrial applications.
Catalog No. 50-276-8359 Supplier Grainger Supplier No. 16T153
The Grainger 16T153 is a single-phase, open drip-proof electric motor with a 1/3 horsepower rating. It has a 115/230 volt input and 1725 RPM speed. The motor is totally enclosed and fan cooled with a 48 frame size. It has a 1.15 service factor and is CSA certified. The 16T153 can be used for various light industrial applications.