Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502763858
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 3 inches and it has an American National Standard Stub Acme thread. This durable tool is made in the USA and well-suited for production threading applications.
Catalog No. 50-276-3858 Supplier Grainger Supplier No. 16T098
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 3 inches and it has an American National Standard Stub Acme thread. This durable tool is made in the USA and well-suited for production threading applications.