Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502764011
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can thread up to 5/8 inch depth of thread. The overall tool length is 3 inches. This single-point threading tool allows both external and internal unified right-hand threads to be accurately cut on lathes, screw machines, and CNC machines. It features TiN coating for increased tool life and superior finishes. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-4011 Supplier Grainger Supplier No. 16T094
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can thread up to 5/8 inch depth of thread. The overall tool length is 3 inches. This single-point threading tool allows both external and internal unified right-hand threads to be accurately cut on lathes, screw machines, and CNC machines. It features TiN coating for increased tool life and superior finishes. The Micro 100 threading tool is made in the USA.