Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502764875
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can thread up to 5/8 inch depth of thread. The overall tool length is 3 inches. This single point threading tool allows longitudinal and face threading applications. It has a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-4875 Supplier Grainger Supplier No. 16T304
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 0.040 inch minimum minor diameter and can thread up to 5/8 inch depth of thread. The overall tool length is 3 inches. This single point threading tool allows longitudinal and face threading applications. It has a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.