Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502766489
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree threading point. It has an overall length of 3-1/2 inches and can cut unified coarse right hand threads from 4-40 to 1/4-20 and unified fine right hand threads from 3-48 to 10-32. The tool has an AlTiN coating for increased tool life and lubricity. It is made in the USA by Micro 100 Tool Corp. This threading tool allows efficient threading operations on CNC lathes.
Catalog No. 50-276-6489 Supplier Grainger Supplier No. 16T295
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree threading point. It has an overall length of 3-1/2 inches and can cut unified coarse right hand threads from 4-40 to 1/4-20 and unified fine right hand threads from 3-48 to 10-32. The tool has an AlTiN coating for increased tool life and lubricity. It is made in the USA by Micro 100 Tool Corp. This threading tool allows efficient threading operations on CNC lathes.