Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765142
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 1/4 inch maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse right hand threads from 4-56 to 1/4-20 TPI. The overall tool length is 3 inches and it weighs 0.04 lbs. This durable threading tool is made in the USA and can be used for both internal and external threading applications.
Catalog No. 50-276-5142 Supplier Grainger Supplier No. 16T152
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 1/4 inch maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse right hand threads from 4-56 to 1/4-20 TPI. The overall tool length is 3 inches and it weighs 0.04 lbs. This durable threading tool is made in the USA and can be used for both internal and external threading applications.