Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765505
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 1/4 inch minimum minor diameter and can cut up to 8 threads per inch. The overall tool length is 3 inches and it has a TiN coating for wear resistance. The threading tool has a right hand cut direction and is made in the USA by Micro 100. It can be used on lathes, screw machines, and CNC machines for both internal and external threading applications. The tool comes with a 30 day money back guarantee.
Catalog No. 50-276-5505 Supplier Grainger Supplier No. 16T291
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a 1/4 inch minimum minor diameter and can cut up to 8 threads per inch. The overall tool length is 3 inches and it has a TiN coating for wear resistance. The threading tool has a right hand cut direction and is made in the USA by Micro 100. It can be used on lathes, screw machines, and CNC machines for both internal and external threading applications. The tool comes with a 30 day money back guarantee.