Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765629
Encompass_Preferred
The Grainger 16T162 is a 1/4 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, sleeve bearing electric motor. It has a 1/2" diameter shaft and measures 4-1/2" in diameter and 5-3/4" in length. This motor can be used for general purpose applications and features thermal overload protection. The enclosure helps protect against dirt and debris.
Catalog No. 50-276-5629 Supplier Grainger Supplier No. 16T162
The Grainger 16T162 is a 1/4 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, sleeve bearing electric motor. It has a 1/2" diameter shaft and measures 4-1/2" in diameter and 5-3/4" in length. This motor can be used for general purpose applications and features thermal overload protection. The enclosure helps protect against dirt and debris.