Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502768303
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a right hand thread direction and can cut unified coarse threads from 4-56 to 1/4-20. The overall length is 3 inches and it has an American National thread form. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-8303 Supplier Grainger Supplier No. 16T429
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree included angle. It has a right hand thread direction and can cut unified coarse threads from 4-56 to 1/4-20. The overall length is 3 inches and it has an American National thread form. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.