Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502768477
Encompass_Preferred
The Grainger 16T322 is a 3/8 in. drive 12-point standard impact socket made of chrome alloy steel with a black phosphate finish. It has a 1-1/16 in. hex size to fit 1-1/16 in. hex fasteners. The impact socket has a precision broached hexagon for accurate fastener fit. It meets ASME B107.300 and is manufactured in Taiwan. The socket has a lifetime warranty. This standard impact socket is designed for use with impact wrenches in heavy-duty applications.
Catalog No. 50-276-8477 Supplier Grainger Supplier No. 16T322
The Grainger 16T322 is a 3/8 in. drive 12-point standard impact socket made of chrome alloy steel with a black phosphate finish. It has a 1-1/16 in. hex size to fit 1-1/16 in. hex fasteners. The impact socket has a precision broached hexagon for accurate fastener fit. It meets ASME B107.300 and is manufactured in Taiwan. The socket has a lifetime warranty. This standard impact socket is designed for use with impact wrenches in heavy-duty applications.