Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502768648
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified coarse right hand threads. The overall length is 3-1/2" and it has a 5/16" minor diameter. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features precision ground thread forms for accurate threading. The Micro 100 brand offers consistent tool life and performance.
Catalog No. 50-276-8648 Supplier Grainger Supplier No. 16T328
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified coarse right hand threads. The overall length is 3-1/2" and it has a 5/16" minor diameter. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features precision ground thread forms for accurate threading. The Micro 100 brand offers consistent tool life and performance.