Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502767200
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point angle and can cut a variety of thread types including UN, UNF, UNEF, UNS, and more. The tool has an overall length of 5" and features TiN coating for increased tool life. It is made in the USA and well-suited for production threading applications. The tool can machine a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics, and more. It is sold individually.
Catalog No. 50-276-7200 Supplier Grainger Supplier No. 16T282
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point angle and can cut a variety of thread types including UN, UNF, UNEF, UNS, and more. The tool has an overall length of 5" and features TiN coating for increased tool life. It is made in the USA and well-suited for production threading applications. The tool can machine a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics, and more. It is sold individually.