Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765336
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The overall length is 3 inches and it has an American National Standard stub length of 2-1/2 inches. The tool is made of M2 high speed steel for strength and tool life. It can machine a variety of materials including steel, cast iron, aluminum, brass, bronze and plastics. The 3/16 inch shank allows it to be used in tool holders accepting that size shank.
Catalog No. 50-276-5336 Supplier Grainger Supplier No. 16T316
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The overall length is 3 inches and it has an American National Standard stub length of 2-1/2 inches. The tool is made of M2 high speed steel for strength and tool life. It can machine a variety of materials including steel, cast iron, aluminum, brass, bronze and plastics. The 3/16 inch shank allows it to be used in tool holders accepting that size shank.