Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502768546
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified coarse right hand threads. The overall length is 5" and it has a TiN coating for increased tool life. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It is made in the USA.
Catalog No. 50-276-8546 Supplier Grainger Supplier No. 16T283
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified coarse right hand threads. The overall length is 5" and it has a TiN coating for increased tool life. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It is made in the USA.