Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502768645
Encompass_Preferred
The Grainger Approved Threading Tool is a high-speed steel threading tool with a 60 degree cutting angle. It has a 1/2" shank diameter and can cut threads from 4 to 56 threads per inch. The overall tool length is 3-1/2". This single-point threading tool allows you to cut accurate internal and external right-hand threads in metal, wood, and plastic. It is designed for use on engine lathes, turret lathes, screw machines, and other machine tools that accept a 1/2" shank tool. The Grainger Approved brand provides industrial supplies of trusted quality at a great value.
Catalog No. 50-276-8645 Supplier Grainger Supplier No. 16T320
The Grainger Approved Threading Tool is a high-speed steel threading tool with a 60 degree cutting angle. It has a 1/2" shank diameter and can cut threads from 4 to 56 threads per inch. The overall tool length is 3-1/2". This single-point threading tool allows you to cut accurate internal and external right-hand threads in metal, wood, and plastic. It is designed for use on engine lathes, turret lathes, screw machines, and other machine tools that accept a 1/2" shank tool. The Grainger Approved brand provides industrial supplies of trusted quality at a great value.