Learn More
Grainger Threading Tool 3/8 Carbide
Supplier:  Grainger 16T386
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree included angle and can cut unified thread forms. The overall length is 3-1/2" and it has a TiN coating for increased tool life. This threading tool is good for both right and left hand internal threading operations. The part number for this tool is 16T386.
Catalog No. 50-276-7252