Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502767253
Encompass_Preferred
The Grainger 3/8" Carbide Threading Tool is a high-speed steel threading tool designed for cutting external right-hand threads. It features a 60 degree threading point with a flat on the side and back for clearance. The overall length is 5" and the shank diameter is 3/8". This threading tool is made of durable carbide that provides good wear resistance and allows faster speeds than high speed steel. It can thread a wide range of materials including steel, stainless steel, cast iron, aluminum, brass, bronze and some plastics. The 3/8" shank fits standard tool holders. This is a high quality threading tool well-suited for production threading applications.
Catalog No. 50-276-7253 Supplier Grainger Supplier No. 16T387
The Grainger 3/8" Carbide Threading Tool is a high-speed steel threading tool designed for cutting external right-hand threads. It features a 60 degree threading point with a flat on the side and back for clearance. The overall length is 5" and the shank diameter is 3/8". This threading tool is made of durable carbide that provides good wear resistance and allows faster speeds than high speed steel. It can thread a wide range of materials including steel, stainless steel, cast iron, aluminum, brass, bronze and some plastics. The 3/8" shank fits standard tool holders. This is a high quality threading tool well-suited for production threading applications.