Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502764895
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right-hand threads. The overall tool length is 3-1/2" and it has a 5/8" minor diameter. This single-point threading tool allows efficient threading operations on lathes. The tool is made of M2 high speed steel for strength and durability. It has a TiN coating for increased hardness and lubricity. The Micro 100 threading tool can be used for both external and internal threading applications.
Catalog No. 50-276-4895 Supplier Grainger Supplier No. 16T126
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right-hand threads. The overall tool length is 3-1/2" and it has a 5/8" minor diameter. This single-point threading tool allows efficient threading operations on lathes. The tool is made of M2 high speed steel for strength and durability. It has a TiN coating for increased hardness and lubricity. The Micro 100 threading tool can be used for both external and internal threading applications.