Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502765983
Encompass_Preferred
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 3/8" shank diameter and can cut unified thread sizes from 4-56 to 1"-8. The inserts are replaceable carbide inserts with a TiN coating and have a screw clamping system for quick changeovers. This is a high quality threading tool well suited for production threading applications where frequent insert changes are required. The replaceable carbide inserts provide a long tool life while the TiN coating reduces friction and heat. The 3/8" shank fits industry standard tool holders.
Catalog No. 50-276-5983 Supplier Grainger Supplier No. 16T376
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 3/8" shank diameter and can cut unified thread sizes from 4-56 to 1"-8. The inserts are replaceable carbide inserts with a TiN coating and have a screw clamping system for quick changeovers. This is a high quality threading tool well suited for production threading applications where frequent insert changes are required. The replaceable carbide inserts provide a long tool life while the TiN coating reduces friction and heat. The 3/8" shank fits industry standard tool holders.