Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502768629
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16ER ANSI standard thread. It has a 60 degree included angle and can cut unified screw threads from 4-48 to 1"-8. The overall length is 5" and it weighs 0.19 lbs. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features TiN coating for increased tool life. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-8629 Supplier Grainger Supplier No. 16T191
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16ER ANSI standard thread. It has a 60 degree included angle and can cut unified screw threads from 4-48 to 1"-8. The overall length is 5" and it weighs 0.19 lbs. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features TiN coating for increased tool life. The Micro 100 threading tool is made in the USA.