Learn More
Grainger Threading Tool 3/8 Carbide
Supplier:  Grainger 16T121
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and is uncoated. The overall length is 5" and it can thread up to 5/8" depth of cut. This is a high quality threading tool made in the USA by Micro 100 for cutting standard unified thread forms. The tool has a precision ground thread profile for accurate threading. It is designed for use on lathes, screw machines, and CNC machines. The 3/8" shank allows it to be held securely in tool holders. This is an essential tool for any machine shop doing threading operations.
Catalog No. 50-276-4874