Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502767358
Encompass_Preferred
The Micro 100 Threading Tool is a 3/8" shank threading tool with a 60 degree threading insert. It has a 0.625" max depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and has an overall length of 5". The replaceable threading insert is made of C6 micrograin carbide for wear resistance and durability. This threading tool allows quick insert changes and can be used for both right and left hand threads. It's well-suited for production threading applications in common materials.
Catalog No. 50-276-7358 Supplier Grainger Supplier No. 16T390
The Micro 100 Threading Tool is a 3/8" shank threading tool with a 60 degree threading insert. It has a 0.625" max depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and has an overall length of 5". The replaceable threading insert is made of C6 micrograin carbide for wear resistance and durability. This threading tool allows quick insert changes and can be used for both right and left hand threads. It's well-suited for production threading applications in common materials.