Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502765181
Encompass_Preferred
The Grainger 16T374 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This compact, portable jack has a welded base for stability and swivel saddle to adjust to any angle. It is designed for use on hard level surfaces and has a built-in safety valve and bypass device. The 16T374 hydraulic bottle jack is ideal for auto maintenance and farm or shop projects requiring up to 16 tons of lifting capacity.
Catalog No. 50-276-5181 Supplier Grainger Supplier No. 16T374
The Grainger 16T374 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This compact, portable jack has a welded base for stability and swivel saddle to adjust to any angle. It is designed for use on hard level surfaces and has a built-in safety valve and bypass device. The 16T374 hydraulic bottle jack is ideal for auto maintenance and farm or shop projects requiring up to 16 tons of lifting capacity.