Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502765391
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right hand threads. The overall tool length is 3-1/2" and it has a 5/8" minor diameter. This threading tool is made in the USA out of C2 grade high speed steel for good wear resistance. It can be used on steels, cast iron, aluminum and other materials. The 3/8" shank allows it to fit industry standard tool holders.
Catalog No. 50-276-5391 Supplier Grainger Supplier No. 16T124
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point and can cut unified coarse right hand threads. The overall tool length is 3-1/2" and it has a 5/8" minor diameter. This threading tool is made in the USA out of C2 grade high speed steel for good wear resistance. It can be used on steels, cast iron, aluminum and other materials. The 3/8" shank allows it to fit industry standard tool holders.