Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502765408
Encompass_Preferred
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 60 degree threading insert that can cut unified, metric, ACME, buttress, and worm threads. The tool has an overall length of 4" and a shank diameter of 3/8". It is designed for light to medium duty threading operations in materials like aluminum, brass, bronze, and plastics. The replaceable threading insert allows for quick insert changes when worn or damaged. This is a high quality threading tool well suited for production threading applications.
Catalog No. 50-276-5408 Supplier Grainger Supplier No. 16T185
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 60 degree threading insert that can cut unified, metric, ACME, buttress, and worm threads. The tool has an overall length of 4" and a shank diameter of 3/8". It is designed for light to medium duty threading operations in materials like aluminum, brass, bronze, and plastics. The replaceable threading insert allows for quick insert changes when worn or damaged. This is a high quality threading tool well suited for production threading applications.