Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502765899
Encompass_Preferred
The Micro 100 3/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 3/8" shank diameter and is 5-1/2" overall length. The insert has a .040" nose radius and can cut unified thread sizes from 4-56 to 1/4-20, metric thread sizes from M1.6 to M6, and pipe thread sizes from 1/16" to 1/2". The tool comes with one DGT060204 threading insert. It is made in the USA.
Catalog No. 50-276-5899 Supplier Grainger Supplier No. 16T382
The Micro 100 3/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 3/8" shank diameter and is 5-1/2" overall length. The insert has a .040" nose radius and can cut unified thread sizes from 4-56 to 1/4-20, metric thread sizes from M1.6 to M6, and pipe thread sizes from 1/16" to 1/2". The tool comes with one DGT060204 threading insert. It is made in the USA.