Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502766083
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point angle and can cut a variety of thread types including UN, UNF, UNEF, UNS, and more. The tool has an overall length of 5" and a flute length of 1-1/2". It is made in the USA out of M2 high speed steel for strength and tool life. The threading tool allows both right and left hand threading.
Catalog No. 50-276-6083 Supplier Grainger Supplier No. 16T128
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has a 60 degree threading point angle and can cut a variety of thread types including UN, UNF, UNEF, UNS, and more. The tool has an overall length of 5" and a flute length of 1-1/2". It is made in the USA out of M2 high speed steel for strength and tool life. The threading tool allows both right and left hand threading.