Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502766092
Encompass_Preferred
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 60 degree threading insert that can cut unified, metric, ACME, buttress, and worm threads. The tool has an overall length of 4" and a shank diameter of 3/8". It is designed for light to medium duty threading operations in materials like aluminum, brass, bronze, and some steels. The tool comes with one threading insert which provides versatility for both internal and external threading operations.
Catalog No. 50-276-6092 Supplier Grainger Supplier No. 16T248
The Micro 100 3/8" shank threading tool is a high speed steel insert style threading tool with a TiN coating for wear resistance. It has a 60 degree threading insert that can cut unified, metric, ACME, buttress, and worm threads. The tool has an overall length of 4" and a shank diameter of 3/8". It is designed for light to medium duty threading operations in materials like aluminum, brass, bronze, and some steels. The tool comes with one threading insert which provides versatility for both internal and external threading operations.