Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502768608
Encompass_Preferred
The Micro 100 Threading Tool is a 3/8" shank threading tool with a 60 degree threading insert. It has a 0.625" maximum depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and uses TPGN style inserts. It has a TiN coating for increased tool life. The overall length of the tool is 5.5". This threading tool allows efficient threading operations in a variety of materials.
Catalog No. 50-276-8608 Supplier Grainger Supplier No. 16T240
The Micro 100 Threading Tool is a 3/8" shank threading tool with a 60 degree threading insert. It has a 0.625" maximum depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and uses TPGN style inserts. It has a TiN coating for increased tool life. The overall length of the tool is 5.5". This threading tool allows efficient threading operations in a variety of materials.