Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502767279
Encompass_Preferred
The Grainger 16T392 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a standard NEMA frame with a C-Face mounting. The motor is designed for general purpose applications.
Catalog No. 50-276-7279 Supplier Grainger Supplier No. 16T392
The Grainger 16T392 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor and is thermally protected. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame. It has a standard NEMA frame with a C-Face mounting. The motor is designed for general purpose applications.