Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502768339
Encompass_Preferred
The Grainger 3/8" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree included angle and 16 threads per inch. The overall length is 5-1/2" and it has a 3/8" shank diameter to fit standard tool holders. Made in the USA, this durable threading tool can cut accurate internal and external unified threads in a variety of materials.
Catalog No. 50-276-8339 Supplier Grainger Supplier No. 16T189
The Grainger 3/8" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree included angle and 16 threads per inch. The overall length is 5-1/2" and it has a 3/8" shank diameter to fit standard tool holders. Made in the USA, this durable threading tool can cut accurate internal and external unified threads in a variety of materials.