Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502768466
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 1/2" maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse, unified fine, and 8 pitch threads. The overall tool length is 3-1/2". This single point threading tool allows quick threading starts and smooth thread finishes in most materials. It is made in the USA.
Catalog No. 50-276-8466 Supplier Grainger Supplier No. 16T252
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 1/2" maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse, unified fine, and 8 pitch threads. The overall tool length is 3-1/2". This single point threading tool allows quick threading starts and smooth thread finishes in most materials. It is made in the USA.