Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502764003
Encompass_Preferred
The Grainger 3/8" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree threading point with a flat on the side and has an overall length of 5". The shank diameter is 3/8" and it can cut unified coarse right-hand threads from 4 to 56 TPI. This durable threading tool is made in the USA and can be used in lathes, screw machines, and other machine tools. The carbide tip provides wear resistance for long tool life.
Catalog No. 50-276-4003 Supplier Grainger Supplier No. 16T066
The Grainger 3/8" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree threading point with a flat on the side and has an overall length of 5". The shank diameter is 3/8" and it can cut unified coarse right-hand threads from 4 to 56 TPI. This durable threading tool is made in the USA and can be used in lathes, screw machines, and other machine tools. The carbide tip provides wear resistance for long tool life.