Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502767038
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has an overall length of 5-1/2" and a cutting diameter of 1/4". This single-point threading tool can be used for both external and internal threading applications. The tool features a TiN coating for increased tool life and lubricity. It is made in the USA out of M2 high-speed steel for good wear resistance. The Micro 100 threading tool is ideal for producing accurate threads in production machining applications.
Catalog No. 50-276-7038 Supplier Grainger Supplier No. 16T125
The Micro 100 Threading Tool is a high-speed steel threading tool with a 3/8" shank diameter and 16 TPI. It has an overall length of 5-1/2" and a cutting diameter of 1/4". This single-point threading tool can be used for both external and internal threading applications. The tool features a TiN coating for increased tool life and lubricity. It is made in the USA out of M2 high-speed steel for good wear resistance. The Micro 100 threading tool is ideal for producing accurate threads in production machining applications.