Learn More
Grainger Threading Tool 3/8 Carbide
Catalog No. 502763976
Encompass_Preferred
The Micro 100 3/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 3/8" shank diameter and is 5-1/2" overall length. The insert has a .040" nose radius and can cut unified thread sizes from 4-56 to 1"-8. It has an American National thread form and comes with one DCGX 432 threading insert. This threading tool allows quick insert changes and reduced downtime. It is rigid and versatile for cutting a wide range of thread sizes.
Catalog No. 50-276-3976 Supplier Grainger Supplier No. 16T056
The Micro 100 3/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 3/8" shank diameter and is 5-1/2" overall length. The insert has a .040" nose radius and can cut unified thread sizes from 4-56 to 1"-8. It has an American National thread form and comes with one DCGX 432 threading insert. This threading tool allows quick insert changes and reduced downtime. It is rigid and versatile for cutting a wide range of thread sizes.