Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502765005
Encompass_Preferred
The Scientific Cutting Tools 5/16-24 Port Thread Port Tool is a high-speed steel port thread cutting tool designed for use on cylinder heads. It features a 5/16-24 UNF thread size and cuts at 500 RPM with SAE #30 motor oil. This 1/4 inch shank tool has an overall length of 5 inches and weighs 0.05 lbs. It is made in the USA out of C2 grade high-speed steel for durability and performance. The port thread cutting tool creates high quality threads in cast iron and aluminum cylinder heads for spark plug ports.
Catalog No. 50-276-5005 Supplier Grainger Supplier No. 16T112
The Scientific Cutting Tools 5/16-24 Port Thread Port Tool is a high-speed steel port thread cutting tool designed for use on cylinder heads. It features a 5/16-24 UNF thread size and cuts at 500 RPM with SAE #30 motor oil. This 1/4 inch shank tool has an overall length of 5 inches and weighs 0.05 lbs. It is made in the USA out of C2 grade high-speed steel for durability and performance. The port thread cutting tool creates high quality threads in cast iron and aluminum cylinder heads for spark plug ports.