Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502766767
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 60 degree included angle. It has a 0.040" minimum minor diameter and can thread up to 5/8" depth of thread. The overall length is 3-1/2" and it has a TiN coating for increased tool life and lubricity. This threading tool is made in the USA and can be used for both right-hand and left-hand internal threading operations. It is sold individually.
Catalog No. 50-276-6767 Supplier Grainger Supplier No. 16T359
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 60 degree included angle. It has a 0.040" minimum minor diameter and can thread up to 5/8" depth of thread. The overall length is 3-1/2" and it has a TiN coating for increased tool life and lubricity. This threading tool is made in the USA and can be used for both right-hand and left-hand internal threading operations. It is sold individually.