Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502766772
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 5/16" shank diameter and an overall length of 4". It has a 60 degree threading point and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool has an AlTiN coating for wear resistance and lubricity. It is made in the USA by Micro 100 and is well-suited for production threading applications on lathes, screw machines, and CNC machines. The tool provides accurate threading with optimized chip flow.
Catalog No. 50-276-6772 Supplier Grainger Supplier No. 16T356
The Micro 100 Threading Tool is a high speed steel threading tool with a 5/16" shank diameter and an overall length of 4". It has a 60 degree threading point and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool has an AlTiN coating for wear resistance and lubricity. It is made in the USA by Micro 100 and is well-suited for production threading applications on lathes, screw machines, and CNC machines. The tool provides accurate threading with optimized chip flow.