Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502767325
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 16 TPI. It has a 60 degree included angle and can cut both right-hand and left-hand threads. The overall length is 3-1/2" and it has a 1/4" max depth of thread. This single-point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-7325 Supplier Grainger Supplier No. 16T113
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 16 TPI. It has a 60 degree included angle and can cut both right-hand and left-hand threads. The overall length is 3-1/2" and it has a 1/4" max depth of thread. This single-point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.