Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502767588
Encompass_Preferred
The Micro 100 1/4" Shank Threading Tool is a high speed steel threading tool with a 1/4" shank diameter and 60 degree threading point. It has an overall length of 4-1/2" and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool has an AlTiN nano coating for increased tool life and lubricity. It is made in the USA by Micro 100, a manufacturer of indexable tooling, solid carbide, and high speed steel cutting tools. This threading tool allows efficient internal and external threading operations on CNC lathes and screw machines.
Catalog No. 50-276-7588 Supplier Grainger Supplier No. 16T357
The Micro 100 1/4" Shank Threading Tool is a high speed steel threading tool with a 1/4" shank diameter and 60 degree threading point. It has an overall length of 4-1/2" and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool has an AlTiN nano coating for increased tool life and lubricity. It is made in the USA by Micro 100, a manufacturer of indexable tooling, solid carbide, and high speed steel cutting tools. This threading tool allows efficient internal and external threading operations on CNC lathes and screw machines.