Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502768453
Encompass_Preferred
The Grainger 5/16"-18 internal threading tap is made of high speed steel with a bright finish. It has a taper length of 3/4", an overall length of 1-1/4", and a spiral point style. This tap is designed for hand tapping applications in low carbon steels, cast iron, brass, aluminum and plastics. It has a chamfered entering end to guide the tap and start the thread. The bright finish provides lubricity. Overall this is a standard 5/16"-18 internal threading tap suitable for common hand tapping applications.
Catalog No. 50-276-8453 Supplier Grainger Supplier No. 16T370
The Grainger 5/16"-18 internal threading tap is made of high speed steel with a bright finish. It has a taper length of 3/4", an overall length of 1-1/4", and a spiral point style. This tap is designed for hand tapping applications in low carbon steels, cast iron, brass, aluminum and plastics. It has a chamfered entering end to guide the tap and start the thread. The bright finish provides lubricity. Overall this is a standard 5/16"-18 internal threading tap suitable for common hand tapping applications.